ENA457509523 Enamine

2-fluoro-4-[(trifluoromethyl)sulfanyl]aniline

  • MDL number MFCD21807292
  • Cas Number : 596850-17-8
  • Molecular Weight : 211.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-219753

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P361,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-219753
pictograms GHS06
hstatements H302,H311,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart