ENA457509551 Enamine

tert-butyl 2,2-dimethyl-1,2-dihydroquinoline-1-carboxylate

  • MDL number MFCD09264185
  • Cas Number : 179898-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219851
Availability details for
Quantity
  • Availability
  • Add to Cart