ENA457509555 Enamine

ethyl 2-[(1-hydroxy-2,3-dihydro-1H-inden-4-yl)oxy]propanoate

  • MDL number MFCD18921255
  • Cas Number : 1494524-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-219869
Availability details for
Quantity
  • Availability
  • Add to Cart