ENA457509566 Enamine

2-(pyrrolidin-3-yloxy)pyridine-4-carbonitrile

  • MDL number MFCD16713970
  • Cas Number : 1249696-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219896
Availability details for
Quantity
  • Availability
  • Add to Cart