ENA457509600 Enamine

1-[5-(phenoxymethyl)-1,2,4-oxadiazol-3-yl]ethan-1-amine

  • MDL number MFCD14625735
  • Cas Number : 1251227-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219163
Availability details for
Quantity
  • Availability
  • Add to Cart