ENA457509615 Enamine

ethyl 5,5-dimethyl-4-oxooxolane-3-carboxylate

  • MDL number MFCD26131224
  • Cas Number : 1803608-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-220020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220020
Availability details for
Quantity
  • Availability
  • Add to Cart