ENA457509637 Enamine

1-(1-phenylethyl)imidazolidin-2-one

  • MDL number MFCD23696385
  • Cas Number : 1547046-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-220067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220067
Availability details for
Quantity
  • Availability
  • Add to Cart