ENA457509873 Enamine

1-methyl-2-(tributylstannyl)-1H-imidazole

  • MDL number MFCD01319030
  • Cas Number : 105494-69-7
  • Molecular Weight : 371.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-220587

Safety Information

signalword Danger
pcodes P260,P264,P270,P273,P280,P301+P310,P302+P352,P305+P351+P338,P312,P314,P330,P332+P313,P337+P313,P362,P363,P391,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-220587
pictograms GHS06,GHS08,GHS09
hstatements H301,H312,H315,H319,H372,H410
Availability details for
Quantity
  • Availability
  • Add to Cart