ENA457509899 Enamine

methyl 2-(2,5-dichlorophenyl)-2-oxoacetate

  • MDL number MFCD26057462
  • Cas Number : 1565067-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-220685

Safety Information

msds https://www.enaminestore.com/catalog/EN300-220685
Availability details for
Quantity
  • Availability
  • Add to Cart