ENA457509960 Enamine

5-methyl-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD20483560
  • Cas Number : 197908-31-9
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-220801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220801
Availability details for
Quantity
  • Availability
  • Add to Cart