ENA457510046 Enamine

{[2-(propylcarbamoyl)ethyl]carbamoyl}formic acid

  • MDL number MFCD16170108
  • Cas Number : 1248664-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-220972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220972
Availability details for
Quantity
  • Availability
  • Add to Cart