ENA457510124 Enamine

(5-fluoro-1H-1,3-benzodiazol-2-yl)(phenyl)methanol

  • MDL number MFCD11106907
  • Cas Number : 33060-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219368
Availability details for
Quantity
  • Availability
  • Add to Cart