ENA457510137 Enamine

5-methyl-1-[(3-methyl-1,2-oxazol-5-yl)methyl]-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD16674352
  • Cas Number : 1248957-71-2
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-221219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221219
Availability details for
Quantity
  • Availability
  • Add to Cart