ENA457510138 Enamine

[(4-chlorophenyl)methyl](methoxy)amine

  • MDL number MFCD21100693
  • Cas Number : 54615-16-6
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-219372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219372
Availability details for
Quantity
  • Availability
  • Add to Cart