ENA457510161 Enamine

2-amino-N-cyclopentyl-N-ethylpentanamide hydrochloride

  • MDL number MFCD28954419
  • Cas Number : 1803570-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-221274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221274
Availability details for
Quantity
  • Availability
  • Add to Cart