ENA457510607 Enamine

N-[(5-nitro-1,3-benzoxazol-2-yl)methylidene]hydroxylamine

  • Cas Number : 27412-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-221710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221710
Availability details for
Quantity
  • Availability
  • Add to Cart