ENA457510662 Enamine

rac-(1R,2R)-2-(4-phenylpiperazine-1-carbonyl)cyclopropane-1-carboxylic acid

  • Cas Number : 1820571-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-222487
Availability details for
Quantity
  • Availability
  • Add to Cart