ENA457510684 Enamine

4-cyclopropanecarbonyl-5,5-dimethyl-2-phenyl-1,2,4-triazolidine-3-thione

  • MDL number MFCD28968350
  • Cas Number : 1803592-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-222866
Availability details for
Quantity
  • Availability
  • Add to Cart