ENA457510966 Enamine

6-chloro-N-ethyl-N-phenylpyrimidin-4-amine

  • MDL number MFCD14611137
  • Cas Number : 405931-48-8
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-222364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222364
Availability details for
Quantity
  • Availability
  • Add to Cart