ENA457511090 Enamine

2-isocyanato-2-phenylpropanenitrile

  • MDL number MFCD30323034
  • Cas Number : 52161-42-9
  • Molecular Weight : 172.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-222737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222737
Availability details for
Quantity
  • Availability
  • Add to Cart