ENA457511108 Enamine

2-amino-2-(1-methoxycyclobutyl)acetic acid hydrochloride

  • MDL number MFCD28968341
  • Cas Number : 1803571-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-222782

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222782
Availability details for
Quantity
  • Availability
  • Add to Cart