ENA457511128 Enamine

1,4-dimethyl-1H-imidazol-5-amine hydrochloride

  • MDL number MFCD23703525
  • Cas Number : 1427475-33-9
  • Molecular Weight : 147.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-222826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222826
Availability details for
Quantity
  • Availability
  • Add to Cart