ENA457511144 Enamine

ethyl 2-[(3-chloro-4-fluorophenyl)sulfanyl]acetate

  • MDL number MFCD11641794
  • Cas Number : 1096907-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-222847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222847
Availability details for
Quantity
  • Availability
  • Add to Cart