ENA457511197 Enamine

5-(4-nitrophenoxy)naphthalen-1-amine

  • MDL number MFCD07343726
  • Cas Number : 26196-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-223058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223058
Availability details for
Quantity
  • Availability
  • Add to Cart