ENA457511201 Enamine

1-(2-methoxyphenyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD11037159
  • Cas Number : 36263-56-6
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-223062

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223062
Availability details for
Quantity
  • Availability
  • Add to Cart