ENA457511252 Enamine

N4-(2-methylpropyl)quinoline-3,4-diamine

  • MDL number MFCD07782115
  • Cas Number : 99010-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-223196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223196
Availability details for
Quantity
  • Availability
  • Add to Cart