ENA457511324 Enamine

tert-butyl 2-methyl-1H,2H,3H-pyrrolo[3,2-c]pyridine-1-carboxylate

  • MDL number MFCD24474482
  • Cas Number : 257937-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-221455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221455
Availability details for
Quantity
  • Availability
  • Add to Cart