ENA457511375 Enamine

2-(4-ethylphenoxy)-N-[1-(1-phenylethyl)piperidin-4-yl]acetamide

  • MDL number MFCD28968252
  • Cas Number : 1687763-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-221899
Availability details for
Quantity
  • Availability
  • Add to Cart