ENA457511483 Enamine

N-(3-amino-4-chlorophenyl)morpholine-4-sulfonamide

  • MDL number MFCD11171582
  • Cas Number : 1042812-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-222173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222173
Availability details for
Quantity
  • Availability
  • Add to Cart