ENA457511512 Enamine

2-(prop-2-enamido)acetic acid

  • MDL number MFCD00085223
  • Cas Number : 24599-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-222214
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart