ENA457511754 Enamine

4-cyanobenzene-1-sulfonyl fluoride

  • MDL number MFCD22691153
  • Cas Number : 33719-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-92
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-224405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-224405
Availability details for
Quantity
  • Availability
  • Add to Cart