ENA457511885 Enamine

cyclopropanethiol

  • MDL number MFCD20233353
  • Cas Number : 6863-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83168

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P370+P378,P391,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-83168
pictograms GHS02,GHS07,GHS09
hstatements H225,H302,H312,H315,H319,H335,H400
Availability details for
Quantity
  • Availability
  • Add to Cart