ENA458424339 Enamine

3-cyclopropyl-3-methylbutanamide

  • MDL number MFCD29033788
  • Cas Number : 1820683-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215575
Availability details for
Quantity
  • Availability
  • Add to Cart