ENA458424349 Enamine

6-[(5-methylpyridin-2-yl)amino]pyridine-2-carbonitrile

  • MDL number MFCD29033798
  • Cas Number : 1820620-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-217542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217542
Availability details for
Quantity
  • Availability
  • Add to Cart