ENA458424354 Enamine

5-chloro-4-cyclopropylpyridin-2-amine

  • MDL number MFCD29033805
  • Cas Number : 1820665-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-217989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217989
Availability details for
Quantity
  • Availability
  • Add to Cart