ENA458424356 Enamine

methyl 2-methyl-1-(propan-2-yl)-1H-pyrrole-3-carboxylate

  • MDL number MFCD29033807
  • Cas Number : 224180-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-218144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-218144
Availability details for
Quantity
  • Availability
  • Add to Cart