ENA458424364 Enamine

2-fluoro-2-(pyrrolidin-3-ylidene)ethan-1-ol

  • Cas Number : 1820749-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-219849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219849
Availability details for
Quantity
  • Availability
  • Add to Cart