ENA458424367 Enamine

3-cyclopropyl-3-methylbutanoyl chloride

  • MDL number MFCD29033831
  • Cas Number : 1021939-07-0
  • Molecular Weight : 160.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-220580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220580
Availability details for
Quantity
  • Availability
  • Add to Cart