ENA458424368 Enamine

ethyl 5,8-difluoroquinoline-3-carboxylate

  • MDL number MFCD29033833
  • Cas Number : 1820608-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-220638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220638
Availability details for
Quantity
  • Availability
  • Add to Cart