ENA458424376 Enamine

2-methyl-3-oxo-2H,3H,5H,6H,7H,8H-imidazo[1,5-a]pyridine-1-carboxylic acid

  • MDL number MFCD29033846
  • Cas Number : 1820641-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-221156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221156
Availability details for
Quantity
  • Availability
  • Add to Cart