ENA458424378 Enamine

1-{4-[1-(hydroxyimino)ethyl]phenyl}pyrrolidine-2,5-dione

  • Cas Number : 1955564-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-221512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221512
Availability details for
Quantity
  • Availability
  • Add to Cart