ENA458424391 Enamine

2,2,2-trifluoroethyl 5-(2,2,2-trifluoroethoxy)pyridine-2-carboxylate

  • MDL number MFCD29033868
  • Cas Number : 1820712-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-222552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-222552
Availability details for
Quantity
  • Availability
  • Add to Cart