ENA458424458 Enamine

4-bromo-1-(2,2-diethoxyethyl)-1H-pyrazole

  • MDL number MFCD08558462
  • Cas Number : 1006682-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118609
Availability details for
Quantity
  • Availability
  • Add to Cart