ENA458424541 Enamine

(1-ethyl-1H-pyrazol-4-yl)methanol

  • MDL number MFCD02253733
  • Cas Number : 905307-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135820
Availability details for
Quantity
  • Availability
  • Add to Cart