ENA458424717 Enamine

2-methyl-1,3-oxazole

  • MDL number MFCD08669512
  • Cas Number : 23012-10-4
  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-176874

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P305+P351+P338,P310,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-176874
pictograms GHS02,GHS05
hstatements H226,H318
Availability details for
Quantity
  • Availability
  • Add to Cart