ENA458424848 Enamine

2-(3,4-dichlorophenoxy)acetonitrile

  • MDL number MFCD00464956
  • Cas Number : 38949-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-201668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201668
Availability details for
Quantity
  • Availability
  • Add to Cart