ENA458424853 Enamine

methyl 2,6-difluoropyridine-4-carboxylate

  • MDL number MFCD14698039
  • Cas Number : 119308-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-202284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202284
Availability details for
Quantity
  • Availability
  • Add to Cart