ENA458425072 Enamine

ethyl 2-[2,6-difluoro-4-(hydroxymethyl)phenoxy]propanoate

  • MDL number MFCD24777600
  • Cas Number : 1502447-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-220209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220209
Availability details for
Quantity
  • Availability
  • Add to Cart