ENA458425079 Enamine

5-bromo-3-[2-(trimethylsilyl)ethynyl]pyridin-2-ol

  • MDL number MFCD26127506
  • Cas Number : 942589-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-220556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-220556
Availability details for
Quantity
  • Availability
  • Add to Cart