ENA458425106 Enamine

N-(2,3-dihydroxypropyl)prop-2-enamide

  • MDL number MFCD19219774
  • Cas Number : 42521-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-221207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221207
Availability details for
Quantity
  • Availability
  • Add to Cart